Learn More
(4R)-3-[[(1S,2S)-2-Hydroxy-3-cyclopenten-1-yl]carbonyl]-4-(phenylmethyl)-2-oxazolidinone, TRC
Specifications
Specifications
| Chemical Name or Material | (4R)-3-(((1S,2S)-2-Hydroxy-3-cyclopenten-1-yl)carbonyl)-4-(phenylmethyl)-2-oxazolidinone |
| CAS | 324741-99-3 |
| Molecular Formula | C16 H17 N O4 |
| Quantity | 500 mg |
| Synonym | (4R)-3-[((1S,2S)-2-Hydroxy-3-cyclopenten-1-yl)carbonyl]-4-(phenylmethyl)-1,3-oxazolidin-2-one |
| InChI Formula | InChI=1S/C16H17NO4/c18-14-8-4-7-13(14)15(19)17-12(10-21-16(17)20)9-11-5-2-1-3-6-11/h1-6,8,12-14,18H,7,9-10H2/t12-,13+,14+/m1/s1 |
| SMILES | O[C@H]1C=CC[C@@H]1C(=O)N2[C@H](Cc3ccccc3)COC2=O |
| Molecular Weight (g/mol) | 287.31 |
| Formula Weight | 287.116 |