accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31972-44-8

H3COPOHNCH3CH3OH3CSCH3OO

CAS RN 31972-44-8

IUPAC Name: [ethoxy(4-methanesulfonyl-3-methylphenoxy)phosphoryl](propan-2-yl)amine
Synonyms: [ethoxy(4-methanesulfonyl-3-methylphenoxy)phosphoryl](isopropyl)amine
Molecular Weight (g/mol): 335.35
Molecular Formula: C13H22NO5PS
InChi Key: LVNYJXIBJFXIRZ-UHFFFAOYNA-N
SMILES: CCOP(=O)(NC(C)C)OC1=CC=C(C(C)=C1)S(C)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)