CAS RN 31972-43-7
CAS RN 31972-43-7
IUPAC Name:
[ethoxy(4-methanesulfinyl-3-methylphenoxy)phosphoryl](propan-2-yl)amine
Synonyms:
[ethoxy(4-methanesulfinyl-3-methylphenoxy)phosphoryl](isopropyl)amine
Molecular Weight (g/mol):
319.36
Molecular Formula:
C13H22NO4PS
InChi Key:
LUQMWGMGWJEGAT-UHFFFAOYNA-N
SMILES:
CCOP(=O)(NC(C)C)OC1=CC=C(C(C)=C1)S(C)=O
1
–
3
of
3
results
1
Fenamiphos Sulfoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fenamiphos Sulfoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fenamiphos Sulfoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.