Learn More
Fenamiphos Sulfoxide, TRC
Specifications
Specifications
| Chemical Name or Material | Fenamiphos-sulfoxide |
| CAS | 31972-43-7 |
| Molecular Formula | C13 H22 N O4 P S |
| Quantity | 50 mg |
| Synonym | Phosphoramidic acid, N-(1-methylethyl)-, ethyl 3-methyl-4-(methylsulfinyl)phenyl ester, Phosphoramidic acid, (1-methylethyl)-, ethyl 3-methyl-4-(methylsulfinyl)phenyl ester (9CI), Phosphoramidic acid, isopropyl-, ethyl 4-(ethylsulfinyl)-m-tolyl ester (8CI), Bay 68138 sulfoxide, Fenamiphos sulfoxide, O-Ethyl O-[4-(methylsulfinyl)-m-tolyl]isopropylphosphoramidate, Fenamiphos-sulfoxide |
| InChI Formula | InChI=1S/C13H22NO4PS/c1-6-17-19(15,14-10(2)3)18-12-7-8-13(20(5)16)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15) |
| SMILES | CCOP(=O)(NC(C)C)Oc1ccc(c(C)c1)S(=O)C |
| IUPAC Name | N-[ethoxy-(3-methyl-4-methylsulfinylphenoxy)phosphoryl]propan-2-amine |
| Molecular Weight (g/mol) | 319.36 |
| Formula Weight | 319.1007 |