CAS RN 31603-00-6
CAS RN 31603-00-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4’-O-Benzyl Oxyphenbutazone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4’-O-Benzyl Oxyphenbutazone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4’-O-Benzyl Oxyphenbutazone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.