Learn More
4’-O-Benzyl Oxyphenbutazone, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-O-Benzyl Oxyphenbutazone |
| CAS | 31603-00-6 |
| Molecular Formula | C26 H26 N2 O3 |
| Quantity | 25 mg |
| Synonym | 4-Butyl-2-phenyl-1-[4-(phenylmethoxy)phenyl]-3,5-pyrazolidinedione, 1-[p-(Benzyloxy)phenyl]-4-butyl-2-phenyl-3,5-pyrazolidinedione |
| InChI Formula | InChI=1S/C26H26N2O3/c1-2-3-14-24-25(29)27(21-12-8-5-9-13-21)28(26(24)30)22-15-17-23(18-16-22)31-19-20-10-6-4-7-11-20/h4-13,15-18,24H,2-3,14,19H2,1H3 |
| SMILES | CCCCC1C(=O)N(N(C1=O)c2ccc(OCc3ccccc3)cc2)c4ccccc4 |
| IUPAC Name | 4-butyl-1-phenyl-2-(4-phenylmethoxyphenyl)pyrazolidine-3,5-dione |
| Molecular Weight (g/mol) | 414.5 |
| Formula Weight | 414.1943 |