CAS RN 316-42-7
CAS RN 316-42-7
IUPAC Name:
dihydrogen (1R)-1-{[(2S,3R,11bS)-3-ethyl-9,10-dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinolin-2-yl]methyl}-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline dichloride
Synonyms:
emetine hydrochloride
Molecular Weight (g/mol):
553.57
Molecular Formula:
C29H42Cl2N2O4
InChi Key:
JROGBPMEKVAPEH-GXGBFOEMSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CC[C@H]1CN2CCC3=CC(OC)=C(OC)C=C3[C@@H]2C[C@@H]1C[C@H]1NCCC2=CC(OC)=C(OC)C=C12
1
–
3
of
3
results
1
Emetine Dihydrochloride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Tocris Bioscience™ Emetine dihydrochloride
RNA polymerase inhibitor; inhibits replication of a broad range of RNA and DNA viruses, including SARS-CoV-2 and Zika
3
Emetine Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.