accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 316-41-6

H3COOONOH3CH3COOONOH3COSOOO

CAS RN 316-41-6

IUPAC Name: bis(16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium) sulfate
Synonyms: bis(berberine) sulfate
Molecular Weight (g/mol): 768.79
Molecular Formula: C40H36N2O12S
InChi Key: OJVABJMSSDUECT-UHFFFAOYSA-L
SMILES: [O-]S([O-])(=O)=O.COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC.COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results