accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 315698-07-8

CH3NNNNOHCl

CAS RN 315698-07-8

IUPAC Name: 5-chloro-7-[(4-ethylpiperazin-1-yl)(pyridin-3-yl)methyl]quinolin-8-ol
Synonyms: 5-chloro-7-[(4-ethylpiperazin-1-yl)(pyridin-3-yl)methyl]quinolin-8-ol
Molecular Weight (g/mol): 382.89
Molecular Formula: C21H23ClN4O
InChi Key: XAFSFXYJOJWLJD-UHFFFAOYNA-N
SMILES: CCN1CCN(CC1)C(C1=CN=CC=C1)C1=CC(Cl)=C2C=CC=NC2=C1O
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)