accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 315-18-4

H3CNHOOCH3NCH3H3CH3C

CAS RN 315-18-4

IUPAC Name: 4-(dimethylamino)-3,5-dimethylphenyl N-methylcarbamate
Synonyms: mexacarbate
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: YNEVBPNZHBAYOA-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC(C)=C(N(C)C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results