accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3147-76-0

H3CCH3H3COHNNN

CAS RN 3147-76-0

IUPAC Name: 2-(2H-1,2,3-benzotriazol-2-yl)-4-tert-butylphenol
Synonyms: 2-(1,2,3-benzotriazol-2-yl)-4-tert-butylphenol
Molecular Weight (g/mol): 267.33
Molecular Formula: C16H17N3O
InChi Key: WXHVQMGINBSVAY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results