Learn More
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole |
| CAS | 3147-76-0 |
| Molecular Formula | C16H17N3O |
| Quantity | 5 g |
| Synonym | 2-(2-Hydroxy-5-tert-butylphenyl)benzotriazole |
| InChI Formula | InChI=1S/C16H17N3O/c1-16(2,3)11-8-9-15(20)14(10-11)19-17-12-6-4-5-7-13(12)18-19/h4-10,20H,1-3H3 |
| SMILES | CC(C)(C)c1ccc(O)c(c1)n2nc3ccccc3n2 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4-tert-butylphenol |
| Molecular Weight (g/mol) | 267.33 |
| Formula Weight | 267.1372 |