CAS RN 31365-76-1
CAS RN 31365-76-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.