Learn More
7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide |
| CAS | 31365-76-1 |
| Molecular Formula | C8H7ClN2OS |
| Quantity | 10 mg |
| Synonym | 4H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1-oxide (8CI, ACI), 7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-oxide |
| InChI Formula | InChI=1S/C8H7ClN2OS/c1-5-10-7-3-2-6(9)4-8(7)13(12)11-5/h2-4H,1H3,(H,10,11) |
| SMILES | CC1=Nc2ccc(Cl)cc2S(=O)N1 |
| IUPAC Name | 7-chloro-3-methyl-2H-1λ^{4},2,4-benzothiadiazine 1-oxide |
| Molecular Weight (g/mol) | 214.67 |
| Formula Weight | 213.9968 |