CAS RN 313348-29-7
CAS RN 313348-29-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-(6-Amino-9-Beta-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(6-Amino-9-b-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(6-Amino-9-Beta-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.