Learn More
1-(6-Amino-9-b-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(6-Amino-9-beta-D-ribofuranosyl-9H-purin-2-yl)-1H-pyrazole-4-carboxylic acid |
| CAS | 313348-29-7 |
| Molecular Formula | C14H15N7O6 |
| Quantity | 100 mg |
| Synonym | Regadenoson Impurity |
| SMILES | Nc1nc(nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)n4cc(cn4)C(=O)O |
| Molecular Weight (g/mol) | 377.31 |
| Formula Weight | 377.1084 |