CAS RN 31251-41-9
CAS RN 31251-41-9
IUPAC Name:
13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-one
Synonyms:
13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-one
Molecular Weight (g/mol):
243.69
Molecular Formula:
C14H10ClNO
InChi Key:
WMQNOYVVLMIZDV-UHFFFAOYSA-N
SMILES:
ClC1=CC2=C(C=C1)C(=O)C1=C(CC2)C=CC=N1
1
–
4
of
4
results
1
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
8-Chloro-5,6-dihydro-11H-benzo[5.6]cyclohepta[1,2-b]pyridin-11-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.