CAS RN 31127-80-7
CAS RN 31127-80-7
IUPAC Name:
N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
Synonyms:
N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol):
747.06
Molecular Formula:
C16H20I3N3O7
InChi Key:
BHCBLTRDEYPMFZ-UHFFFAOYNA-N
SMILES:
CC(=O)NC1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
1
–
4
of
4
results
1
5-(Acetylamino)-N,N’-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(Acetylamino)-N,N’-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
5-(Acetylamino)-N,N’-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.