CAS RN 3112-74-1
CAS RN 3112-74-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Copper Dipropanoate, TRC
CAS: 3112-74-1 Molecular Formula: C6H10CuO4 Molecular Weight (g/mol): 209.69 Synonym: Copper Dipropionate,Copper(2+) Propionate,Copper(II) Propanoate,Copper(II) Propionate,Cupric Propanoate,Cupric Propionate IUPAC Name: copper;propanoate SMILES: [Cu+2].CCC(=O)[O-].CCC(=O)[O-]