accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 311-45-5

H3COPOOCH3ONOO

CAS RN 311-45-5

IUPAC Name: diethyl 4-nitrophenyl phosphate
Synonyms: paraoxon ethyl paraoxon diethyl paraoxon phosphacol
Molecular Weight (g/mol): 275.2
Molecular Formula: C10H14NO6P
InChi Key: WYMSBXTXOHUIGT-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results