Learn More
Paraoxon, TRC
Specifications
Specifications
| Chemical Name or Material | Paraoxon-ethyl |
| CAS | 311-45-5 |
| Molecular Formula | C10 H14 N O6 P |
| Quantity | 250 mg |
| Synonym | Paraoxon Ethyl, Paraoxon |
| InChI Formula | InChI=1S/C10H14NO6P/c1-3-15-18(14,16-4-2)17-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3 |
| SMILES | CCOP(=O)(OCC)Oc1ccc(cc1)[N+](=O)[O-] |
| IUPAC Name | diethyl (4-nitrophenyl) phosphate |
| Molecular Weight (g/mol) | 275.2 |
| Formula Weight | 275.0559 |