accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 311-09-1

ClClH3CNCH3HNOONHNCH3H3C

CAS RN 311-09-1

IUPAC Name: benzyl({3-[(4-{[3-(benzyldiethylazaniumyl)propyl]amino}-3,6-dioxocyclohexa-1,4-dien-1-yl)amino]propyl})diethylazanium dichloride
Synonyms: benzoquinonium chloride
Molecular Weight (g/mol): 617.7
Molecular Formula: C34H50Cl2N4O2
InChi Key: WQRJZLXQCYZJEU-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].CC[N+](CC)(CCCNC1=CC(=O)C(NCCC[N+](CC)(CC)CC2=CC=CC=C2)=CC1=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results