accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3102-70-3

CH3CH3NH2H3CH2N

CAS RN 3102-70-3

IUPAC Name: 2,4,6-trimethylbenzene-1,3-diamine
Synonyms: 2,4,6-trimethylbenzene-1,3-diamine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: ZVDSMYGTJDFNHN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results