Learn More
2,4,6-Trimethyl-1,3-benzenediamine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trimethyl-1,3-benzenediamine |
| CAS | 3102-70-3 |
| Molecular Formula | C9H14N2 |
| Quantity | 1 g |
| Synonym | 2,4-Mesitylenediamine, 1,3,5-Trimethyl-2,6-benzenediamine, 2,4,6-Trimethyl-1,3-benzenediamine, 2,4,6-Trimethyl-1,3-phenylenediamine, 2,4,6-Trimethyl-m-phenylenediamine, NSC 10392 |
| InChI Formula | InChI=1S/C9H14N2/c1-5-4-6(2)9(11)7(3)8(5)10/h4H,10-11H2,1-3H3 |
| SMILES | Cc1cc(C)c(N)c(C)c1N |
| IUPAC Name | 2,4,6-trimethylbenzene-1,3-diamine |
| Molecular Weight (g/mol) | 150.22 |
| Formula Weight | 150.1157 |