CAS RN 309923-18-0
CAS RN 309923-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione, TRC
CAS: 309923-18-0 Molecular Formula: C7 H12 N4 O2 Molecular Weight (g/mol): 184.1958 Synonym: Terbuthylazine Metabolite CGA 324007,1,3,5-Triazine-2,4(1H,3H)-dione, 6-[(1,1-dimethylethyl)amino]-,6-[(1,1-Dimethylethyl)amino]-1,3,5-triazine-2,4(1H,3H)-dione,LM5,Terbuthylazine Metabolite LM5 IUPAC Name: 6-(tert-butylamino)-1,3,5-triazine-2,4-diol SMILES: CC(C)(C)Nc1nc(O)nc(O)n1
2
Terbuthylazine metabolite CGA 324007 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality