accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 305849-16-5

CH3H3CSOOHNFFFCH3

CAS RN 305849-16-5

IUPAC Name: 2,4,6-trimethyl-N-{[4-(trifluoromethyl)phenyl]methyl}benzene-1-sulfonamide
Synonyms: 2,4,6-trimethyl-N-{[4-(trifluoromethyl)phenyl]methyl}benzenesulfonamide
Molecular Weight (g/mol): 357.39
Molecular Formula: C17H18F3NO2S
InChi Key: FVZQXGUIEZGZJJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NCC1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results