accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FHOFN

CAS RN 2967-54-6

IUPAC Name: 3,5-difluoro-4-hydroxybenzonitrile
Synonyms: 3,5-difluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: XZNZJDPPWWFJAL-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=C1F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results