accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSNNH3C

CAS RN 2882-20-4

IUPAC Name: 2-methyl-3-(methylsulfanyl)pyrazine
Synonyms: methyl-methylthiopyrazine
Molecular Weight (g/mol): 140.2
Molecular Formula: C6H8N2S
InChi Key: PPPFFGVGWFKTHX-UHFFFAOYSA-N
SMILES: CSC1=NC=CN=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results