accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOH3CNHOO

CAS RN 28819-05-8

IUPAC Name: methyl 2-{[(benzyloxy)carbonyl]amino}propanoate
Synonyms: methyl 2-{[(benzyloxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: OMDVFTVXPVXANK-UHFFFAOYNA-N
SMILES: COC(=O)C(C)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results