CAS RN 2872-48-2
CAS RN 2872-48-2
IUPAC Name:
1,4-diamino-2-methoxy-9,10-dihydroanthracene-9,10-dione
Synonyms:
disperse red 11
Molecular Weight (g/mol):
268.27
Molecular Formula:
C15H12N2O3
InChi Key:
TUXJTJITXCHUEL-UHFFFAOYSA-N
SMILES:
COC1=CC(N)=C2C(=O)C3=CC=CC=C3C(=O)C2=C1N
1
–
1
of
1
results
1
Disperse Red 11 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality