accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClOF

CAS RN 286474-59-7

IUPAC Name: 6-chloro-2-fluoro-3-methylbenzaldehyde
Synonyms: 6-chloro-2-fluoro-3-methylbenzaldehyde
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: GGZOMBUJFMOEEZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results