accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2831-66-5

ClNNClN

CAS RN 2831-66-5

IUPAC Name: 2,4-dichloro-1,3,5-triazine
Synonyms: 2,4-dichloro-1,3,5-triazine
Molecular Weight (g/mol): 149.96
Molecular Formula: C3HCl2N3
InChi Key: OMRXVBREYFZQHU-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=NC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results