CAS RN 27876-24-0
CAS RN 27876-24-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hexylpyridine, TRC
CAS: 27876-24-0 Molecular Formula: C11 H17 N Molecular Weight (g/mol): 163.259 IUPAC Name: 4-hexylpyridine SMILES: CCCCCCc1ccncc1