accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH

CAS RN 2777-66-4

IUPAC Name: methyl undec-10-ynoate
Synonyms: methyl undec-10-ynoate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: TXFNJLOAXFOJGK-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results