accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 276695-22-8

NNHOHNClClHCl

CAS RN 276695-22-8

IUPAC Name: 8,9-dichloro-1H,2H,3H,4H,4aH,5H,6H-pyrazino[1,2-a]quinoxalin-5-one hydrochloride
Synonyms: 8,9-dichloro-1H,2H,3H,4H,4aH,6H-pyrazino[1,2-a]quinoxalin-5-one hydrochloride
Molecular Weight (g/mol): 308.59
Molecular Formula: C11H12Cl3N3O
InChi Key: YPNWSZJDAKOUAW-UHFFFAOYNA-N
SMILES: Cl.ClC1=C(Cl)C=C2N3CCNCC3C(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results