CAS RN 27668-52-6
CAS RN 27668-52-6
IUPAC Name:
dimethyl(octadecyl)[3-(trimethoxysilyl)propyl]azanium chloride
Synonyms:
dimethyl(octadecyl)[3-(trimethoxysilyl)propyl]azanium chloride
Molecular Weight (g/mol):
496.29
Molecular Formula:
C26H58ClNO3Si
InChi Key:
WSFMFXQNYPNYGG-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC
1
–
2
of
2
results
1
Promotions Available
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol