Learn More
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium Chloride (40-50 wt. % in methanol), TRC
Specifications
Specifications
| CAS | 27668-52-6 |
| Molecular Formula | C26H58NO3Si.Cl |
| Quantity | 1 mL |
| Synonym | (3-(Trimethoxysilyl)propyl)octadecyldimethylammonium chloride3-(Trimethoxysilyl)propyl-N-octadecyl-N,N-dimethylammonium chloride5700 Antimicrobial AgentA 9-6346AEM 5700AEM 5700 AntimicrobialAY 43-006Aegis AEM 5700AgrishieldBioShield 7200BioShield AM 500BioShield AM 500IBiosafe MCX 117683C 18NTMSCA 200CA 200 (quaternary compd.)DA 5700DC 5700DK-Q 8-8905DMAOPDMAOP (onium compound)DMOAPDow Corning 5701Dow Corning 9-6346Goldshield 5HM 98-3LS 6985MonoFoilMonoFoil Antimicrobial LiquidN-Octadecyldimethyl[3-(trimethoxysilyl)propyl]ammonium chlorideOctadecyldimethyl(trimethoxysilyl)propylammonium chlorideOctadecyldimethyl[3-(trimethoxysilyl)propyl]ammonium chloridePC 5700Polon MF 50ProShield 5000DQ 9-5700Q 9-6346Quat-SilsesquioxaneSIO 6620SiQACXS 70-241XZ 2-2300[3-(Trimethoxysilyl)propyl]dimethyloctadecylammonium chloride |
| InChI Formula | InChI=1S/C26H58NO3Si.ClH/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-24-27(2,3)25-23-26-31(28-4,29-5)30-6;/h7-26H2,1-6H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC |
| IUPAC Name | dimethyl-octadecyl-(3-trimethoxysilylpropyl)azanium;chloride |
| Molecular Weight (g/mol) | 496.3 |
| Formula Weight | 495.3874 |
| Concentration or Composition (by Analyte or Components) | Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium Chloride 40-50% |
| Show More |