CAS RN 2761281-51-8
CAS RN 2761281-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ PROTAC™(H-PGDS)-8
Tocris Bioscience™ PROTAC™(H-PGDS)-8
Negative control for PROTAC™(H-PGDS)-7 (Cat. No. 8004)