CAS RN 27546-07-2
IUPAC Name:
dioxomolybdenumdiol
Synonyms:
molybdic acid H2MoO4 molybdic(VI) acid
Molecular Weight (g/mol):
161.96
Molecular Formula:
H2MoO4
InChi Key:
VLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILES:
O[Mo](O)(=O)=O
1
–
3
of
3
results
1
Ammonium molybdate (di), 4% w/v aq. soln.
CAS: 27546-07-2 Molecular Formula: H2MoO4 Molecular Weight (g/mol): 161.96 MDL Number: MFCD00010882 InChI Key: VLAPMBHFAWRUQP-UHFFFAOYSA-L
2
Ammonium molybdate (di), 10% w/v aq. soln.
CAS: 27546-07-2 Molecular Formula: H2MoO4 Molecular Weight (g/mol): 161.96 MDL Number: MFCD00010882 InChI Key: VLAPMBHFAWRUQP-UHFFFAOYSA-L
3
Ammonium molybdate (di), Mo 56.5%
CAS: 27546-07-2 Molecular Formula: H2MoO4 Molecular Weight (g/mol): 161.96 MDL Number: MFCD00010882 InChI Key: VLAPMBHFAWRUQP-UHFFFAOYSA-L Synonym: Ammonium dimolybdate