accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2747-04-8

H3COOOOBr

CAS RN 2747-04-8

IUPAC Name: 4-(bromomethyl)-2-oxo-2H-chromen-7-yl acetate
Synonyms: 4-(bromomethyl)-2-oxochromen-7-yl acetate
Molecular Weight (g/mol): 297.1
Molecular Formula: C12H9BrO4
InChi Key: YDJFMNSSDOXBSR-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C2C(CBr)=CC(=O)OC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results