Learn More
4-Bromomethyl-7-acetoxycoumarin, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromomethyl-7-Acetoxycoumarin |
| CAS | 2747-04-8 |
| Molecular Formula | C12 H9 Br O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C12H9BrO4/c1-7(14)16-9-2-3-10-8(6-13)4-12(15)17-11(10)5-9/h2-5H,6H2,1H3 |
| SMILES | CC(=O)Oc1ccc2C(=CC(=O)Oc2c1)CBr |
| IUPAC Name | [4-(bromomethyl)-2-oxochromen-7-yl] acetate |
| Molecular Weight (g/mol) | 297.1 |
| Formula Weight | 295.9684 |