CAS RN 2741956-55-6
CAS RN 2741956-55-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ BAY 390
Tocris Bioscience™ BAY 390
Potent and selective TRPA1 antagonist. Brain penetrant and orally bioavailable