Learn More
Tocris Bioscience™ BAY 390
Product Code. 30058518
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30058518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058518 Supplier Tocris Bioscience™ Supplier No. 7978/10
Please to purchase this item. Need a web account? Register with us today!
Potent and selective TRPA1 antagonist. Brain penetrant and orally bioavailable
BAY 390 is a potent and selective transient receptor potential ankyrin 1 (TRPA1) antagonist (IC50 values are 16, 63 and 82 nM for hTRPA1 FLIPR, rTRPA1 FLIPR and hTRPA1 Ephys, respectively). In vivo, BAY 390 exhibits efficacy in rat models of inflammatory and neuropathic pain. BAY 390 is orally bioavailable and brain penetrant. Suitable to use as an in vitro or in vivo probe to investigate the roles of TRPA1 in models of acute, inflammatory, and neuropathic pain.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R,2R)-2-[(4-Fluorophenyl)amino]-1-(trifluoromethyl)cyclohexanol
CAS 2741956-55-6
Quantity 10 mg
Target TRPA1 Blockers
Molecular Formula C13H15F4NO
Purity 0.98