CAS RN 2727065-34-9
CAS RN 2727065-34-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[3-Acetyl-4-[(2RS)-3-(diethylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.