accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CHNOCl

CAS RN 26930-17-6

IUPAC Name: 4-chloro-N-ethylbenzamide
Synonyms: 4-chloro-N-ethylbenzamide
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: TXRYLYLDCHDRSJ-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results