accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261763-47-7

CH3OHOFF

CAS RN 261763-47-7

IUPAC Name: 2-(2,3-difluoro-4-methylphenyl)acetic acid
Synonyms: (2,3-difluoro-4-methylphenyl)acetic acid
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: XYMLKJRXFGXFCJ-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results