CAS RN 261762-90-7
CAS RN 261762-90-7
IUPAC Name:
2-chloro-6-fluoro-3-methylphenol
Synonyms:
2-chloro-6-fluoro-3-methylphenol
Molecular Weight (g/mol):
160.57
Molecular Formula:
C7H6ClFO
InChi Key:
RPTGCWUNGWVQET-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C(O)=C(F)C=C1
1
–
1
of
1
results
1
2-Chloro-6-fluoro-3-methylphenol, 97%, Thermo Scientific™
CAS: 261762-90-7 Molecular Formula: C7H6ClFO Molecular Weight (g/mol): 160.572 MDL Number: MFCD01631366 InChI Key: RPTGCWUNGWVQET-UHFFFAOYSA-N Synonym: acmc-20anyl,2-chloro-6-fluoro-m-cresol,2-chloro-6-fluoro-3-methyl-phenol,2-chloro-4-fluoro-3-hydroxytoluene PubChem CID: 2773683 IUPAC Name: 2-chloro-6-fluoro-3-methylphenol SMILES: CC1=C(C(=C(C=C1)F)O)Cl