accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

(R)(R)O(S)(R)∗OHOOHOHO∗n

CAS RN 25990-10-7

IUPAC Name: (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
Synonyms: α-D-galacturonic acid α-D-galactopyranuronic acid
Molecular Weight (g/mol): 194.14
Molecular Formula: C6H10O7
InChi Key: AEMOLEFTQBMNLQ-BKBMJHBISA-N
SMILES: O[C@H]1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)C(O)=O
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)