accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClONH

CAS RN 254883-43-7

IUPAC Name: 4-(2-chloro-5-methylphenoxy)piperidine
Synonyms: 4-(2-chloro-5-methylphenoxy)piperidine
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: MTMIZXIWQUAWTF-UHFFFAOYSA-N
SMILES: CC1=CC(OC2CCNCC2)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results