accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CI

CAS RN 25309-64-2

IUPAC Name: 1-ethyl-4-iodobenzene
Synonyms: 1-ethyl-4-iodobenzene
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: OOLSRHZMXAYDFB-UHFFFAOYSA-N
SMILES: CCC1=CC=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results